2,2',4,4',6,6'-Hexachlorobiphenyl (PCB No. 155)
Catalog # EHERC20015500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:360.88 g/mol
EINECS:251-773-8
CAS Number:33979-03-2
Formula:C₁₂H₄Cl₆