Order Entry
Export
ContactUsLinkComponent
2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44)
2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44)
Catalog # EHERC20004400
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,5'-Tetrachlorobiphenyl (PCB No. 44)
Catalog # EHERC20004400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :291.99 g/mol
UN:3432
ADR:9,II
CAS Number:41464-39-5
Formula:C₁₂H₆Cl₄