2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193)
Catalog # PROOCIL-PCB-193-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:395.32 g/mol
CAS Number:69782-91-8
Formula:C₁₂H₃Cl₇