Order Entry
Export
ContactUsLinkComponent
2,2',3,4',5,5',6-Heptachlorobiphenyl (PCB No. 187)
2,2',3,4',5,5',6-Heptachlorobiphenyl (PCB No. 187)
Catalog # EHERC20018700
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,4',5,5',6-Heptachlorobiphenyl (PCB No. 187)
Catalog # EHERC20018700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:52663-68-0
Formula:C₁₂H₃Cl₇