Order Entry
Export
ContactUsLinkComponent
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
Catalog # EHERC20015700
Supplier:  EHRENSTORFER
CAS Number:  
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
Catalog # EHERC20015700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
ADR:9,II
 :360.88 g/mol
UN:3432
CAS Number:69782-90-7
Formula:C₁₂H₄Cl₆