Order Entry
Export
ContactUsLinkComponent
2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191)
2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191)
Catalog # PROOCIL-PCB-191-C
CAS Number:  
2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191)
Catalog # PROOCIL-PCB-191-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:74472-50-7
Formula:C₁₂H₃Cl₇