Order Entry
Export
ContactUsLinkComponent
2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170)
2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170)
Catalog # EHERC20017000
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170)
Catalog # EHERC20017000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
UN:3432
ADR:9,II
CAS Number:35065-30-6
Formula:C₁₂H₃Cl₇