Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)
2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)
2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)
PROOCIL-PCB-160-C
:
LGC Standards PROMOCHEM
:
2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)
PROOCIL-PCB-160-C
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
360.88 g/mol
:
41411-62-5
:
(C₆H₄Cl)(C₆Cl₅)