Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Phenylbenzaldehyde, Sigma-Aldrich®

Irritant
Supplier:   Merck
4-Phenylbenzaldehyde, Sigma-Aldrich®
Supplier:  Merck
Certificates
4-Phenylbenzaldehyde, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
  • CAS Number:3218-36-8
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00006947
  • Molecular Weight:182.22 g/mol
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Refrigerator
  • EINECS:221-742-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 39,90 €
    39,90 €
    1 * 5 g
       39,90 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 122,00 €
    122,00 €
    1 * 25 g
       122,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 222,00 €
    222,00 €
    1 * 100 g
       222,00 €