2,2'-Dibromobiphenyl (PBB No. 4), Sigma-Aldrich®
Catalog # SIAL697885-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:312 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:13029-09-9
Formula:C₁₂H₈Br₂
MDL Number:MFCD00093707