2,4,6-Tris(bromomethyl)mesitylene, Sigma-Aldrich®
Catalog # SIAL388467-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:398.96 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:244-697-1
UN:3261
CAS Number:21988-87-4
Formula:C₁₂H₁₅Br₃
MDL Number:MFCD00192554