Order Entry
Export
Orders LinkContactUsLinkComponent
Mesitylene, Sigma-Aldrich®

Mesitylene, Sigma-Aldrich®

Glass bottle, 2,5 L
Catalog # SIAL140864-2.5L
Flammable
Irritant
Environment
Supplier:   Merck
Mesitylene, Sigma-Aldrich®
Catalog # SIAL140864-2.5L
Supplier:  Merck
CAS Number:  108-67-8
Mesitylene, Sigma-Aldrich®

Glass bottle, 2,5 L

CAS Number:  108-67-8
Price
119,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8637 g/cm³ (20 °C)
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • UN:2325
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-604-4
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications