Order Entry
Export
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)benzyl alcohol, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)benzyl alcohol, Sigma-Aldrich®
Catalog # SIAL263370-5G
Supplier:  Merck
CAS Number:  
3,5-Bis(trifluoromethyl)benzyl alcohol, Sigma-Aldrich®
Catalog # SIAL263370-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :244.14 g/mol
Boiling Pt:100 °C (14 mmHg)
Melting Pt:53…54 °C
 :M-Clarity™ quality level = MQ200
EINECS:251-168-9
CAS Number:32707-89-4
Formula:C₉H₆F₆O
MDL Number:MFCD00009907