
3,5-Bis(trifluoromethyl)benzyl alcohol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32707-89-4
:
Formula:C₉H₆F₆O
MDL Number:MFCD00009907
:244.14 g/mol
Boiling Pt:40 °C (0.5 mmHg)
Melting Pt:52…56 °C
Flash Pt:97 °C (206 °F)
Storage Temperature:Ambient
EINECS:251-168-9
:GHS07