4,4'-Dimethylbiphenyl, Sigma-Aldrich®
Catalog # SIALD151203-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.27 g/mol
Boiling Pt:295 °C (1013 hPa)
Melting Pt:117…122 °C
Density:1.102 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:210-337-7
CAS Number:613-33-2
Formula:C₁₄H₁₄
MDL Number:MFCD00008545