4,4'-Di-tert-butylbiphenyl, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1625-91-8
:
Formula:C₂₀H₂₆
MDL Number:MFCD00008834
Boiling Pt:228 °C (1013 hPa)
Melting Pt:127 °C
Density:0.9691 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:266.43 g/mol
EINECS:216-615-4