Order Entry
Export
ContactUsLinkComponent
4,4'-Dimethylbenzil
4,4'-Dimethylbenzil
Catalog # MOLE15183383-25G
Supplier:  Molekula
CAS Number:  
4,4'-Dimethylbenzil
Catalog # MOLE15183383-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :238.29 g/mol
Boiling Pt:209…211 °C (7 mmHg)
Melting Pt:102…105 °C
EINECS:222-388-2
CAS Number:3457-48-5
Formula:C₁₆H₁₄O₂
MDL Number:MFCD00008554