

4,4'-Bis(dimethylamino)benzophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:90-94-8
:
Formula:C₁₇H₂₀N₂O
MDL Number:MFCD00008312
:268.36 g/mol
Merck Index:12,06265
Boiling Pt:428 °C (1013 hPa)
Melting Pt:172 °C
Flash Pt:220 °C
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:202-027-5
:GHS05
:GHS08
Storage Temperature:Ambient