

4,4'-Bis(dimethylamino)benzophenone 98%
:90-94-8
:
:C₁₇H₂₀N₂O
:MFCD00008312
:268.36 g/mol
:12,06265
:428 °C (1013 hPa)
:171…176 °C
:220 °C
:Ambient
:202-027-5
:6.1,III
:GHS08
:GHS05
- Appearance:Blue to dark blue to grey Powder
- Infrared spectrum:Conforms
- Melting point:171 to 176 °C (Dec)
- HPLC:≥97.5 %
- Titration with HClO4:≥97.5 %