Order Entry
Export
ContactUsLinkComponent
4,4'-Bis(diethylamino)benzophenone 99+%
Irritant
Environment
4,4'-Bis(diethylamino)benzophenone 99+%
Certificates
undefined
Irritant
Environment
4,4'-Bis(diethylamino)benzophenone 99+%

Specification Test Results

  • Appearance:
    White to yellow to yellow-green Powder or granules and/or lumps
  • Infrared spectrum:
    Conforms
  • Melting point:
    93 to 96 °C
  • HPLC:
    ≥99.0 %

Specification Test Results

Related Information