Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylbenzophenone

3-Methylbenzophenone

Glass bottle for solids, 100 mg
Catalog # PROOMM0001.13
3-Methylbenzophenone
Catalog # PROOMM0001.13
CAS Number:  643-65-2
3-Methylbenzophenone

Glass bottle for solids, 100 mg

CAS Number:  643-65-2
Price
900,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  m-Methylbenzophenone
  • CAS Number:643-65-2
  • Formula:C₁₄H₁₂O
  • MDL Number:MFCD00008535
  • Molecular Weight:196.25 g/mol
  • Boiling Pt:317 °C (1013 hPa)
  • Melting Pt:0…2 °C
  • Density:1.088 g/cm³ (17.5 °C)
  • EINECS:211-401-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications