3-Methylbenzophenone
Glass bottle for solids, 100 g
Catalog # MOLEM23534468
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: m-Methylbenzophenone
- CAS Number:643-65-2
- Formula:C₁₄H₁₂O
- MDL Number:MFCD00008535
- Molecular Weight:196.25 g/mol
- Boiling Pt:317 °C (1013 hPa)
- Melting Pt:0…2 °C
- Density:1.088 g/cm³ (17.5 °C)
- EINECS:211-401-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



