Order Entry
Export
ContactUsLinkComponent
1,1-Dichloro-2,2-bis(4-ethylphenyl)ethane
Environment
1,1-Dichloro-2,2-bis(4-ethylphenyl)ethane
Catalog # EHERC16000000
Supplier:  EHRENSTORFER
CAS Number:  
Environment
1,1-Dichloro-2,2-bis(4-ethylphenyl)ethane
Catalog # EHERC16000000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :307.26 g/mol
Boiling Pt:376 °C (1013 hPa)
Melting Pt:59.85 °C
EINECS:200-785-1
UN:3077
ADR:9,III
 :GHS09
CAS Number:72-56-0
Formula:C₁₈H₂₀Cl₂