

p-Xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-42-3
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556
:106.17 g/mol
Merck Index:12,10214
Boiling Pt:138.3 °C (1013 hPa)
Melting Pt:13 °C
Density:1.096 g/cm³ (20 °C)
EINECS:203-396-5
UN:1307
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:25 °C
:GHS02
:GHS07
Storage Temperature:Ambient