

1,2-Diethylbenzene, Sigma-Aldrich®
Catalog # SIALD90601-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
Boiling Pt:183 °C (1013 hPa)
Melting Pt:–31.2 °C
Density:0.8662 g/cm³ (18 °C)
EINECS:205-170-1
UN:2049
ADR:3,III
CAS Number:135-01-3