Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dimethylthiophenol, Sigma-Aldrich®

2,5-Dimethylthiophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL275468-5G
Irritant
Supplier:   Merck
2,5-Dimethylthiophenol, Sigma-Aldrich®
Catalog # SIAL275468-5G
Supplier:  Merck
CAS Number:  4001-61-0
2,5-Dimethylthiophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  4001-61-0
Price
54,10 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,5-Dimethylbenzenethiol, 2,5-Dimethylbenzene-1-thiol, 2-Mercapto-p-xylene, 1,4-Dimethyl-2-mercaptobenzene
  • CAS Number:4001-61-0
  • Formula:C₈H₁₀S
  • MDL Number:MFCD00010020
  • Molecular Weight:138.23 g/mol
  • Boiling Pt:48…49 °C (0.6 mmHg)
  • Density:1.0313 g/cm³ (20 °C)
  • EINECS:223-649-3
  • UN:2810
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications