
2,6-Dimethylthiophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:118-72-9
:
Formula:C₈H₁₀S
MDL Number:MFCD00010021
:138.23 g/mol
Boiling Pt:76 °C (1013 hPa)
Density:1.038 g/cm³ (25 °C)
EINECS:204-272-3
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07