
3,4-Dimethylthiophenol, Sigma-Aldrich®
Catalog # SIAL269689-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:138.23 g/mol
Boiling Pt:58…59 °C (1 mmHg)
Melting Pt:54…56 °C
Density:1.0256 g/cm³ (20 °C)
EINECS:242-587-8
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:18800-53-8
Formula:C₈H₁₀S
MDL Number:MFCD00010023