


Thymol ≥98.5%, Sigma-Aldrich®
:89-83-8
:
:2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
:MFCD00002309
:150.22 g/mol
:13,09476
:232 °C (lit.)
:48…51 °C (lit.)
:0.965 g/cm³ (25 °C) (lit.)
:116 °C (closed cup)
:Room Temperature
:201-944-8
:2430
:8,III
:GHS05
:GHS07
:GHS09
- Appearance (Colour):Colorless to White
- Appearance (Form):Powder or Crystals
- Solubility (Colour):Colorless
- Solubility (Turbidity)
50 mg/ml, EtOH:Clear - 1H NMR Spectrum:Conforms to Structure
- Purity (HPLC):≥98.5 %
- Recommended Retest Period:3 years