2,6-Di-tert-butyl-4-methoxyphenol, Sigma-Aldrich®
SIAL251062-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:236.35 g/mol
:135…140 °C (10 mmHg)
:105.4 °C
:M-Clarity™ quality level = MQ200
:207-693-0
:489-01-0
:C₁₅H₂₄O₂
:MFCD00008824