4-Nitro-m-xylene, Sigma-Aldrich®
Catalog # SIAL171018-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:151.16 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:9 °C
Density:1.135 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-947-4
UN:1665
ADR:6.1,II
:M-Clarity™ quality level = MQ100
CAS Number:89-87-2
Formula:C₈H₉NO₂
MDL Number:MFCD00007169