Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitro-m-xylene, Sigma-Aldrich®

2-Nitro-m-xylene, Sigma-Aldrich®

Supplier:   Merck
2-Nitro-m-xylene, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-Nitro-m-xylene, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  2-Nitro-m-xylene, 1,3-Dimethyl-2-nitrobenzene, 2,6-Dimethylnitrobenzene
  • CAS Number:81-20-9
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00007181
  • Molecular Weight:151.16 g/mol
  • Boiling Pt:∼225 °C (1013 hPa)
  • Melting Pt:14…16 °C
  • Density:1.112 g/cm³ (20 °C)
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • EINECS:201-333-6
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • UN:1665
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 33,90 €
    33,90 €
    1 * 5 g
       33,90 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 65,70 €
    65,70 €
    1 * 100 g
       65,70 €