2-Nitro-4,5-xylidine, Sigma-Aldrich®
Catalog # SIALD172200-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:143 °C
Density:1.398 g/cm³ (–123.15 °C)
EINECS:230-211-5
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:166.18 g/mol
UN:2811
CAS Number:6972-71-0
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD00007811