Order Entry
Export
ContactUsLinkComponent
3,5-Di-tert-butylaniline, Sigma-Aldrich®
Irritant
3,5-Di-tert-butylaniline, Sigma-Aldrich®
Catalog # SIAL273104-1G
Supplier:  Merck
CAS Number:  
Irritant
3,5-Di-tert-butylaniline, Sigma-Aldrich®
Catalog # SIAL273104-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.34
 :M-Clarity™ quality level = MQ100
EINECS:219-173-0
 :GHS07
CAS Number:2380-36-1
Formula:C₁₄H₂₃N
MDL Number:MFCD00134096