Order Entry
Export
ContactUsLinkComponent
2,6-Dimethylphenyl isocyanate, Sigma-Aldrich®
2,6-Dimethylphenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL252336-5G
Supplier:  Merck
CAS Number:  
2,6-Dimethylphenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL252336-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :147.18 g/mol
Boiling Pt:87…80 °C (1013 hPa)
Density:1.057 g/cm³ (20 °C)
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
UN:2206
CAS Number:28556-81-2
Formula:C₉H₉NO
MDL Number:MFCD00002012