
5-Iodo-m-xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:22445-41-6
:
Formula:C₈H₉I
MDL Number:MFCD00060659
:232.06 g/mol
Boiling Pt:234…235 °C (1013 hPa)
Density:1.61 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Refrigerator