
5-Iodo-m-xylene 99%
ACRO309250050
:
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Appearance:Clear light yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5930 to 1.5960 (20°C, 589 nm)
- GC:≥98.5 %
:
:232.06 g/mol
:92…94 °C (3 mmHg)
:1.6 g/cm³
:>110 °C
:GHS07
:Refrigerator
:22445-41-6
:C₈H₉I
:MFCD00060659