
4-Iodo-o-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31599-61-8
:
Formula:C₈H₉I
MDL Number:MFCD00040989
:232.06 g/mol
Boiling Pt:231…233 °C
Density:1.633
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient