0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5-Fluoro-m-xylene
5-Fluoro-m-xylene
5-Fluoro-m-xylene
5-Fluoro-m-xylene
Supplier:
Thermo Scientific
Certificates
Expand image
5-Fluoro-m-xylene
Supplier:
Thermo Scientific
About this item
CAS Number:
461-97-2
:
Formula:
C₈H₉F
MDL Number:
MFCD00052366
:
124.16 g/mol
Boiling Pt:
145 °C (1013 hPa)
UN:
1993
ADR:
3,III