
5-Fluoro-m-xylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:461-97-2
:
Formula:C₈H₉F
MDL Number:MFCD00052366
:124.16 g/mol
Boiling Pt:144…145 °C
Density:0.983
Flash Pt:35 °C (95 °F)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02