Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Fluoro-m-xylene

Supplier:   Thermo Scientific
2-Fluoro-m-xylene
Supplier:  Thermo Scientific
Certificates
2-Fluoro-m-xylene

2 Options Available Below

Product Details & Documents
Synonyms:  2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene
  • CAS Number:443-88-9
  • Formula:C₈H₉F
  • MDL Number:MFCD00039215
  • Molecular Weight:124.16 g/mol
  • Boiling Pt:165…167 °C (1013 hPa)
  • Density:0.988 g/cm³ (20 °C)
  • EINECS:207-144-5
  • UN:1993
  • ADR:3,III
  •  :Density: 0.988 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Discontinued
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g