Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethylbenzamide
2,6-Dimethylbenzamide
2,6-Dimethylbenzamide
2,6-Dimethylbenzamide
Supplier:
CHEMPUR
Certificates
Expand image
2,6-Dimethylbenzamide
Supplier:
CHEMPUR
About this item
CAS Number:
55321-98-7
:
Formula:
C₉H₁₁NO
:
149.19 g/mol