4'-Methylacetophenone
1 g
Catalog # ROTH3603.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methyl p-tolyl ketone, p-Methylacetophenone, 1-(p-Tolyl)ethanone
- CAS Number:122-00-9
- Formula:C₉H₁₀O
- MDL Number:MFCD00008751
- Molecular Weight:134.18 g/mol
- Boiling Pt:226 °C (1013 hPa)
- Melting Pt:28 °C
- Density:1.013 g/cm³ (20 °C)
- Flash Pt:92 °C
- Storage Temperature:Ambient
- EINECS:204-514-8
Specifications
Specifications
- Pk:1 g



