2-Chloro-p-xylene, Sigma-Aldrich®
Catalog # SIAL26630-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:140.61 g/mol
Boiling Pt:180…185 °C (1013 hPa)
Melting Pt:1.5 °C
Density:1.06 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-444-2
CAS Number:95-72-7
Formula:C₈H₉Cl
MDL Number:MFCD00000564