
α-Chloro-m-xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:620-19-9
:
Formula:C₈H₉Cl
MDL Number:MFCD00000909
:140.61 g/mol
Merck Index:12,10224
Boiling Pt:196 °C (1013 hPa)
Density:1.064 g/cm³ (20 °C)
EINECS:210-628-9
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
:GHS07
Storage Temperature:Refrigerator