

α,α'-Dichloro-p-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-25-6
:
Formula:C₈H₈Cl₂
MDL Number:MFCD00000920
:175.06 g/mol
Boiling Pt:254 °C
Melting Pt:98…102 °C
Density:1.417
Storage Temperature:Ambient
EINECS:210-782-7
UN:2928
ADR:6.1,II
:GHS06
:GHS05