Order Entry
Export
ContactUsLinkComponent
α,α'-Dichloro-m-xylene, Sigma-Aldrich®
α,α'-Dichloro-m-xylene, Sigma-Aldrich®
Catalog # SIAL36580-25G-F
Supplier:  Merck
CAS Number:  
α,α'-Dichloro-m-xylene, Sigma-Aldrich®
Catalog # SIAL36580-25G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :175.06 g/mol
Boiling Pt:254 °C (1013 hPa)
Melting Pt:33…35 °C
Density:1.302 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:210-932-1
UN:2928
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
CAS Number:626-16-4
Formula:C₈H₈Cl₂
MDL Number:MFCD00000912