Order Entry
Export
ContactUsLinkComponent
α,α'-Dichloro-o-xylene
α,α'-Dichloro-o-xylene
Catalog # MOLE14273214-100G
Supplier:  Molekula
CAS Number:  
α,α'-Dichloro-o-xylene
Catalog # MOLE14273214-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :175.06 g/mol
Boiling Pt:240 °C (1013 hPa)
Melting Pt:54…55 °C
Density:1.393 g/cm³ (0 °C)
EINECS:210-291-8
UN:2928
ADR:6.1,II
CAS Number:612-12-4
Formula:C₈H₈Cl₂
MDL Number:MFCD00000903