Order Entry
Export
ContactUsLinkComponent
 
Irritant
4-Hydroxy-3,5-dimethylbenzaldehyde, Sigma-Aldrich®
Catalog # SIAL140392-5G
Supplier:  Merck
CAS Number:  
Irritant
4-Hydroxy-3,5-dimethylbenzaldehyde, Sigma-Aldrich®
Catalog # SIAL140392-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :150.18 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:218-774-5
 :GHS07
Storage Temperature:Ambient
CAS Number:2233-18-3
Formula:C₉H₁₀O₂
MDL Number:MFCD00006946