
5-Bromo-m-xylene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:556-96-7
:
Formula:C₈H₉Br
MDL Number:MFCD00000087
Boiling Pt:60 - 62 °C (5,33 hPa)
Density:1.36 g/cm³
Flash Pt:87 °C
:185.06 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5475 to 1.5495 (20°C, 589 nm)
- GC:≥97.5 %